Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (DyLight® 800)

Supplier: Rockland Immunochemicals
610-445-002-0.5
RL6104450020.5EA 432 USD InStock
RL6104450020.5
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 800)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty