Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (DyLight® 549)

Supplier: Rockland Immunochemicals
611-142-122 611-142-122
RL611-142-122EA 134 USD InStock
RL611-142-122 CA611-142-122
Anti-IgG Goat Polyclonal Antibody (DyLight® 549)
Antibodies
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty