Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
611-1924 611-1924
RL611-1924EA 270 USD InStock
CA611-1924 RL611-1924
Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgG F(ab')2 Reacts with Rabbit (Lapine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty