Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GABRG2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-C81
RL612-401-C81EA 518 USD InStock
RL612-401-C81
Anti-GABRG2 Rabbit Polyclonal Antibody
Antibodies
Anti-GABA(A) Receptor gamma 2 (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of 46 kDa in size corresponding to the gamma 2 subunit of the GABA(A).

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: GABRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty