Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP2K5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-D74
RL612-401-D74EA 518 USD InStock
RL612-401-D74
Anti-MAP2K5 Rabbit Polyclonal Antibody
Antibodies
Anti-MEK5 pS311/pT315 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 49 kDa in size corresponding to MEK5 Protein phosphorylated at Ser311/Thr315.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAP2K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty