Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRM5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-D77
RL612-401-D77EA 518 USD InStock
RL612-401-D77
Anti-GRM5 Rabbit Polyclonal Antibody
Antibodies
Anti-mGluR5/1a (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 125 kDa monomer and 250 kDa dimer in size corresponding to mGluR5.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500

Type: Primary
Antigen: GRM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty