Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-D92
RL612-401-D92EA 518 USD InStock
RL612-401-D92
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
Anti-NMDA R2B pT1252 Antibody is suitable for use in Western Blotting and IHC. Expect a band of approximately 180 kDa in size corresponding to NMDAR NR2B subunit Protein phosphorylated at Tyr1252 in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:400;

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty