Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPH3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-E21
RL612-401-E21EA 518 USD InStock
RL612-401-E21
Anti-RPH3A Rabbit Polyclonal Antibody
Antibodies
Anti-Rabphilin 3A (Rabbit) anitbody is suitable for use in ELISA, and in western blotting. Expect a band of approximately 82 kDa in size corresponding to rabpihilin 3A Protein in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RPH3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty