Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-E38
RL612-401-E38EA 518 USD InStock
RL612-401-E38
Anti-SYT1 Rabbit Polyclonal Antibody
Antibodies
Anti-Synaptotagmin pT202 (Rabbit) antibody is suitable for use in Western Blotting and IHC. Expect a band of approximately 60-62 kDa in size corresponding to synaptotagmin Protein phosphorylated at Thr202 in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:400

Type: Primary
Antigen: SYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty