Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
614-4504 614-4504
RL614-4504EA 278 USD InStock
RL614-4504 CA614-4504
Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Secondary Rabbit Anti-F(ab')2 Reacts with Pig (Swine, Porcine)

Recommended Dilutions: ELISA: 1:1000 - 1:3000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty