Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
617-103-007 617-103-007
RL617-103-007EA 312 USD InStock
CA617-103-007 RL617-103-007
Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Monkey IgM (mu chain) peroxidase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000-1:100000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty