Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG gamma GoatPolyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
617-105-012 617-105-012
RL617-105-012EA 326 USD InStock
RL617-105-012 CA617-105-012
Anti-IgG gamma GoatPolyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Anti-Monkey IgG alkaline phosphatase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunochemistry as well as other alkaline phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: gamma chain
Host: Goat
Isotype: IgG
Reactivity: Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty