Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgA alpha Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
618-109-006 618-109-006
RL618-109-006EA 195 USD InStock
CA618-109-006 RL618-109-006
Anti-IgA alpha Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgA Reacts with Ferret

Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: alpha chain
Host: Goat
Isotype: IgA
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty