Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG gamma Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
618-109-012 618-109-012
RL618-109-012EA 195 USD InStock
CA618-109-012 RL618-109-012
Anti-IgG gamma Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgG (gamma chain) Reacts with Ferret

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: gamma chain
Host: Goat
Isotype: IgG
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty