Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

F(ab')2 Anti-Biotin Goat Polyclonal Antibody (Cy3.5®)

Supplier: Rockland Immunochemicals
700-112-098 700-112-098
RL700-112-098EA 645 USD InStock
CA700-112-098 RL700-112-098
F(ab')2 Anti-Biotin Goat Polyclonal Antibody (Cy3.5®)
Antibodies
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:100-1:1000; Flow Cytometry: 1:200-1:1400; Flow Cytometry: 1:200-1:1400
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Cy3.5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty