Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals
710-4020 710-4020
RL710-4020EA 220 USD InStock
CA710-4020 RL710-4020
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Secondary Rabbit Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty