Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
711-1524 711-1524
RL711-1524EA 308 USD InStock
RL711-1524 CA711-1524
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
This product has been assayed against 1.0 µg of Rabbit IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty