Would you like to log in again?
Tissue Powders for Adsorption, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
C110 C107 C113 C105 C121 C116 C101 C110 C105 C106 C113 C107 C114 C122 C116 C102 C101 C109 C103 C117 C103 C102 C115 C124 C120 C104 C115 C123 C118 C100 C122 C100 C108 C120 C104 C124 C112 C111 C108 C111 C112 C119 C123 C109 C118 C106 C114 C121 C117 C119
RLC119EA 440 USD InStock
RLC119 RLC114 RLC113 RLC112 RLC111 RLC118 RLC117 RLC116 RLC115 RLC110 CAC103 CAC102 CAC101 RLC109 CAC100 RLC108 RLC103 RLC102 RLC101 CAC109 RLC100 CAC108 RLC107 CAC107 CAC106 RLC106 CAC105 RLC105 CAC104 RLC104 CAC114 CAC113 CAC112 CAC111 CAC110 CAC119 CAC118 CAC117 CAC116 CAC115 CAC124 CAC123 CAC122 CAC121 CAC120 RLC124 RLC123 RLC122 RLC121 RLC120
Tissue Powders for Adsorption, Rockland Immunochemicals
Media for Microbiology Dehydrated Media for Microbiology
Use for protocols that require a high nuclear protein content
Powders are produced by washing the various organs repeatedly in saline to remove blood, precipitated in acetone, and micronized to a fine powder. Keep container sealed and store at 2–8°C (36–46°F). All vials contain 1g of powder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty