Use for protocols that require a high nuclear protein content
Powders are produced by washing the various organs repeatedly in saline to remove blood, precipitated in acetone, and micronized to a fine powder. Keep container sealed and store at 2–8°C (36–46°F). All vials contain 1g of powder.