Would you like to log in again?
Mouse Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
C301-0050 C301-0100 C301-0010
RLC301-0050EA 2309 USD InStock
RLC301-0050 RLC301-0010 RLC301-0100
Mouse Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Mouse Complement is ideal for hemolytic assay where hemolysis of IgG antibodies are required.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty