Would you like to log in again?
Rabbit 'A' (Nesting) Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
C302-0010
RLC302-0010EA 1037 USD InStock
RLC302-0010
Rabbit 'A' (Nesting) Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty