Would you like to log in again?
Rat Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
C305-0050 C305-0100 C305-0010
RLC305-0050EA 1383 USD InStock
RLC305-0050 RLC305-0010 RLC305-0100
Rat Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Rat Complement is suitable for CFT and SRH.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty