Would you like to log in again?
Gamma Globulin Fractions, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D611-0050 D600-0100 D605-0100 D613-1000 D612-1000 D608-0100 D609-0100 D610-1000 D603-0100 D607-1000 D602-0100 D606-0100
RLD611EA 163 USD InStock
RLD611 RLD600 RLD612 RLD613 RLD602 RLD603 RLD610 RLD608 RLD609 RLD605 RLD606 RLD607
Gamma Globulin Fractions, Rockland Immunochemicals
Protein Purification Protein Purification Reagents
Fractions are produced by delipidation, salt fractionation, and ion exchange chromatography followed by extensive dialysis against phosphate buffered saline
They are then filtered through 0.22µm filters and lyophilized. Fractions contain IgG with trace quantities of other immunoglobulins.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty