Would you like to log in again?
Freund's Complete Adjuvant, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D614-0100 D614-0050 D614-0050 D614-0100
RLD614EA 124.63 USD InStock
CAD614-0100 RLD614 10801-490 RLD614-0100
Freund's Complete Adjuvant, Rockland Immunochemicals
Protein Expression Protein Expression Reagents
Freund's Complete Adjuvant (CFA)
  • water-in-oil emulsions with antigens for the injection of immunogens for antibody production
  • Autoclaved at 250°C
Special formulation of adjuvant containing paraffin oil, surfactant and heat-killed Mycobacterium butyricum.
Freund’s adjuvant may be used to form water-in-oil emulsions with antigens for the injection of immunogens for antibody production. Proper emulsion formation is essential and should be tested by observing a drop of the emulsion on the surface of water which should 'bead and not disperse. Antigens properly emulsified stimulate a strong and long-lasting immune response due to the slow release of antigen into the host’s system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty