Would you like to log in again?
Fetal Bovine Serum, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
FBS-02-0050
RLFBS-02-0050EA 139 USD InStock
RLFBS-02-0050
Fetal Bovine Serum, Rockland Immunochemicals
Sera (Serum)
Sterile Fetal Bovine Serum (FBS)
  • 0.1 μm sterile filtered
  • Mycoplasma free
  • Endotoxin <1.0EU/mL
  • Hemoglobin <15mg%
This product originates in the United States and is mycoplasma, virus, performance and endotoxin tested.
Fetal Bovine Serum sourced from the United States is collected exclusively from USDA licensed abattoirs from within the United States. Raw material is processed through 0.1 μm (100nm) filters in compliance with regulatory guidelines. Animals are inspected pre-mortem and post-mortem by USDA veterinarians as free of any infectious diseases. Serum is maintained between 4-6°F after collection through sterile filling to ensure exceptional performance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty