Would you like to log in again?
Chemiluminescent HRP Substrates, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
FEMTOMAX-110 FEMTOMAX-110
RLFEMTOMAX-110EA 475 USD InStock
RLFEMTOMAX-110 CAFEMTOMAX-110
Chemiluminescent HRP Substrates, Rockland Immunochemicals
Electrophoresis Reagents Western Blotting Reagents
Nonradioactive, enhanced luminol-based chemiluminescent HRP substrates are designed for the detection of HRP (horseradish peroxidase) through western blotting and EIA (enzyme immunoassay) use
Substrates are supplied as clear, colorless, odorless liquids. They should be protected from light and stored at 4°C (39°F) prior to opening.
The highly sensitive PicoMax* allows for the detection of picogram (10-12) amounts of antigen using photographic film or other imaging methods. The extremely sensitive FemtoMax* allows for the detection of femtogram (10-15) amounts of antigen using photographic film or other imaging methods, including highly sensitive CCD cameras.
Ordering information: Supplied as two components; one 55mL luminol reagent and one 55mL reaction buffer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty