Would you like to log in again?
ELISA Kit, Antibody And Blocking Solution, Starter Pack, Rockland Immunochemical
Supplier: Rockland Immunochemicals
K-500
RLK-500EA 518 USD InStock
RLK-500
ELISA Kit, Antibody And Blocking Solution, Starter Pack, Rockland Immunochemical
Assays ELISAs
Antibody starter pack contains key reagents necessary to perform ELISA, immunohistochemistry, and immunofluorescence. Included are antibodies suitable for ELISA, immunohistochemistry, immunoprecipitation, immunofluorescent microscopy, and western blotting.
Delivery information: Antibody starter pack contains conjugated secondary antibodies, ECL substrate, blocking solution, and monoclonal Anti-FLAG antibody.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty