Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NF-kB (P65) Transcription Factor Polyclonal Antibody

Supplier: Rockland Immunochemicals
KAA065
RLKAA065EA 930 USD InStock
RLKAA065
Anti-NF-kB (P65) Transcription Factor Polyclonal Antibody
Antibodies
Primary Anti-NF-kB (P65) TRANSCRIPTION FACTOR Reacts with Pan

Type: Primary
Antigen: NF-kB (P65) Transcription Factor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty