Would you like to log in again?
Loading Control Antibody Sampler Kit
Supplier: Rockland Immunochemicals
KAS-001
RLKAS001EA 471 USD InStock
RLKAS001
Loading Control Antibody Sampler Kit
Antibodies
The Loading Control Antibody Sampler Kit contains antibodies to a variety of housekeeping proteins. Loading controls are essential for the interpretation of western blot data. Loading control antibodies allow the verification of equal protein loading between samples. Ideal loading controls are expressed constitutively and at high levels with low variability between cell lines and experimental conditions. Also, the loading control should be at a different molecular weight than the protein of interest this allows the bands to be visually distinguishable. The Loading Control Antibody Sampler kit contains enough primary and secondary antibodies to perform ten western blots per loading control antibody.
Recommended Dilutions: ELISA: 1:5,000 - 1:25,000; Western Blot: 1:500 - 1:5,000;
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty