Would you like to log in again?
Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody, FemtoMax™, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KCA002 KCA002
RLKCA002EA 339 USD InStock
RLKCA002 CAKCA002
Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody, FemtoMax™, Rockland Immunochemicals
Assays Immunological Assays
This Chemiluminescent Western Blot Kit allows for the detection of primary mouse polyclonal or monoclonal antibodies provided by the user
Recommended Dilutions: Western Blot: 1:20,000; Wash buffers MUST NOT contain SODIUM AZIDE or other inhibitors of peroxidase activity!
After protein separation and transfer, the membrane is probed with primary antibody. Detection of the membrane bound primary antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems. Because of the extremely high sensitivity of our FemtoMax Chemiluminescent Substrate Kit for Western Blotting, primary and secondary antibodies can be used at greater dilutions. If you currently dilute your primary antibody 1:1,000 using an ECL substrate equivalent, then dilute your antibody 1:5,000 using the FemtoMax substrate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty