Would you like to log in again?
Tris HCl Buffer Solutions 1.0 M, Rockland
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-002 MB-004 MB-002 MB-001 MB-005 MB-004 MB-001 MB-003 MB-005 MB-003
RLMB-004EA 83 USD InStock
RLMB-004 RLMB-005 RLMB-002 RLMB-003 RLMB-001 CAMB-001 CAMB-002 CAMB-003 CAMB-004 CAMB-005
Tris HCl Buffer Solutions 1.0 M, Rockland
Buffers Biological Buffers
Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
These solutions were aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The products were tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: These products ares for research use only and are not intended for therapeutic or diagnostic applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty