Would you like to log in again?
Buffers, TE solutions 10X concentrates (TRIS-EDTA buffer), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-006 MB-006 MB-007 MB-007
RLMB-006EA 123 USD InStock
CAMB-006 CAMB-007 RLMB-006 RLMB-007
Buffers, TE solutions 10X concentrates (TRIS-EDTA buffer), Rockland Immunochemicals
Buffers Biological Buffers
TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA, cDNA or RNA
TE buffer is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 100 mM Tris HCl and 10 mM EDTA at a pH of 8.0. Meticulously prepared using ultra pure reagents dissolved in DEPC treated molecular biology grade water.
TE is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.
Caution: These products ares for research use only and are not intended for therapeutic or diagnostic applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty