Would you like to log in again?
PBS and TBS buffers, Rockland
Supplier: Rockland Immunochemicals
MB-011 MB-008 MB-011 MB-012 MB-012 MB-008
RLMB-011EA 112 USD InStock
RLMB-011 RLMB-012 CAMB-011 CAMB-012 RLMB-008 CAMB-008
PBS and TBS buffers, Rockland
Buffers Electrophoresis Buffers Washing Buffers
Phosphate buffered and Tris buffered saline are suitable for multiple applications including biological diluent buffer for antibodies or other biologics
Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.
Store container at room temperature (18° to 26° C) prior to opening. If desired, the solution may be stored at 4° C or less. Some salts may precipitate out of solution at lower temperature. Allow buffer to equilibrate to room temperature (18° to 26° C) to restore solubility of some salts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty