Would you like to log in again?
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND
Supplier: Rockland Immunochemicals
MB-014 MB-014
RLMB-014EA 112 USD InStock
RLMB-014 CAMB-014
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND
Buffers Cloning Buffers
EDTA is widely used for scavenging metal ion in biochemistry and molecular biology
In tissue culture EDTA is used as a chelating agent that binds to calcium and prevents joining of cadherins between cells, preventing clumping of cells grown in liquid suspension, or detaching adherent cells for passaging. EDTA is also known to inhibit a range of metallopeptidases, the method of inhibition occurs via the chelation of the metal ion required for catalytic activity.
Ethylenediaminetetraacetic acid is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 0.5 M Trisodium EDTA at pH 8.0. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water (DI) treated with diethyl pyrocarbonate (DEPC).
Ion depletion is commonly used to deactivate metal-dependent enzymes, either as an assay for their reactivity or to suppress damage to DNA or proteins
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty