Would you like to log in again?
SDS-Page Running Gel Buffers, 10X Concentrate
Supplier: Rockland Immunochemicals
MB-017 MB-017
RLMB-017EA 128 USD InStock
CAMB-017 RLMB-017
SDS-Page Running Gel Buffers, 10X Concentrate
Buffers Electrophoresis Buffers Running Buffers
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature
SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins
This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionized water (or equivalent) to its final working concentration. No pH adjustment is required. 10X SDS-PAGE Running Buffer consists of 0.25 M Tris HCl, 1.92 M Glycine and 1% (w/v) Sodium Dodecyl Sulfate (SDS); pH 8.3. Meticulously prepared using ultra-pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty