Would you like to log in again?
GET Buffer, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-043 MB-043
RLMB-043EA 105 USD InStock
CAMB-043 RLMB-043
GET Buffer, Rockland Immunochemicals
Buffers Biological Buffers
GET Buffer pH 8.0, 50 mM Glucose, 10 mM EDTA, 25 mM Tris HCl, pH 8.0 buffer
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty