Would you like to log in again?
SSC Buffer, 20X Concentrate, pH 7.0, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-045 MB-045
RLMB-045EA 150 USD InStock
RLMB-045 CAMB-045
SSC Buffer, 20X Concentrate, pH 7.0, Rockland Immunochemicals
Buffers Electrophoresis Buffers Hybridization Buffers
Saline-sodium citrate (SSC) buffer is most commonly used as a buffer for the hybridization process and washing in experiments where spcies of nucleic or ribonucleic acid are to be matched to complimentary sequences (such as Southern and Nouthern blotting, in situ hybridization, or DNA microarrays)
20X SSC pH 7.0 (3.0 M Sodium Chloride, 0.3 M Sodium Citrate, pH 7.0) DEPC Treated is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water treated with diethyl pyrocarbonate (DEPC).
SSC is used to help control the stringency of hybridization in these steps.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty