Would you like to log in again?
TM Buffer, 1X Tris-Magnesium sulphate, pH 7.5
Supplier: Rockland Immunochemicals
MB-059 MB-059
RLMB-059EA 109 USD InStock
RLMB-059 CAMB-059
TM Buffer, 1X Tris-Magnesium sulphate, pH 7.5
Buffers Biological Buffers
This product is ready-to-use as a working 1X solution and requires no further dilution
This buffer consists of 50 mM Tris HCl, 10 mM magnesium sulphate at pH 7.5 and was prepared using ultra pure reagents dissolved in pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty