Would you like to log in again?
Blocking Buffer, TBS Fish Gel 10X, Rockland
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-067-0100 MB-067-1000 MB-067-0100 MB-067-1000
RLMB-067-0100EA 112 USD InStock
RLMB-067-0100 RLMB-067-1000 CAMB-067-0100 CAMB-067-1000
Blocking Buffer, TBS Fish Gel 10X, Rockland
Buffers Electrophoresis Buffers Blocking Buffers
Highly sensitive ELISA assays require minimal non-specific interactions
This product is a 10X concentrated stock solution. Prepare a 1X working solution by diluting 1 part 10X concentrate with 9 parts distilled-deionized water or equivalent. 10X TBS Fish Gel Concentrate consists of 1.0 M Tris hydrochloride, 1.5 M sodium chloride and Fish Gelatin at pH 7.5. A proprietary combination of stabilizers and preservatives are used that are azide and mercury free.
Non-specific binding results in elevated background levels and a decrease in signal-to-noise ratios. Blocking reagents act to minimize non-specific interactions of secondary reactants with each other and with the primary solid phase binding sites.
This product is a 10X concentrated stock solution. Prepare a 1X working solution by diluting 1 part 10X concentrate with 9 parts distilled-deionized water or equivalent. 10X TBS Fish Gel Concentrate consists of 1.0 M Tris hydrochloride, 1.5 M sodium chloride and Fish Gelatin at pH 7.5. A proprietary combination of stabilizers and preservatives are used that are azide and mercury free.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Storage Condition: Store container at 4° C before opening. Protect from moisture and light. No special shipping conditions or precautions are required.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty