Would you like to log in again?
Blocking Buffer For Immunohistochemistry Serum And Azide Free, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-071-0100
RLMB-071-0100EA 165 USD InStock
RLMB-071-0100
Blocking Buffer For Immunohistochemistry Serum And Azide Free, Rockland Immunochemicals
Buffers Electrophoresis Buffers Blocking Buffers
Protein blocker specifically designed for immunohistochemical staining where detection occurs using an immunoenzymatic reaction with substrate, i.e. peroxidase (HRP) with diaminobenzidine (DAB)
  • Specifically inhibits non-specific staining of IHC detection reagents
  • Undiluted or 1:1 in TBS
  • The buffer is pH adjusted to pH 7.6.
A detailed protocol for immunohistochemical staining is available at www.rockland-inc.com.
Application Note: This immunohistochemistry protein blocker is specifically designed for immunohistochemical staining where detection occurs using an immunoenzymatic reaction with substrate, i.e. peroxidase (HEP) with diaminobenzidine (DAB). This product specifically inhibits non-specific staining of IHC detection reagents. It may be used on various specimens, including cultured cells and primary tissue to lower background staining. Optimum performance is achieved using this product undiluted.  However, Immunohistochemistry Protein Blocker can often be diluted 1:1 in TBS without a significant loss of performance. This immunohistochemistry protein blocker consists of proprietary blocking agents and stabilizers prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty