Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from salmon sperm)

Supplier: Rockland Immunochemical

MB-103-1000 MB-103-0025 MB-103-1000 MB-103-0025
RLMB-103-1000EA 179 USD InStock
CAMB-103-1000 RLMB-103-1000 RLMB-103-0025 CARLMB103-0025
DNA (Deoxyribonucleic acid) (from salmon sperm)
DNA (Deoxyribonucleic acid)
This product is a convenient, ready-to-use solution of salmon sperm DNA especially prepared for use in the preparation of pre-hybridization and hybridization solutions and as a DNA carrier in yeast transformation protocols and other related methods

This solution contains sheared single-stranded DNA molecules that can be used to block the non-specific attachment of probe DNA to the surface of a membrane (Southern) or to increase yeast transformation efficiency.


MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty