Would you like to log in again?
Bovine Ribonuclease A, DNase- and Protease-Free (from Pancreas)
Supplier: Rockland Immunochemicals
MB-113-0005 MB-113-0005 MB-113-0001 MB-113-0001
RLMB-113-0001EA 109 USD InStock
CAMB-113-0005 RLMB-113-0001 RLMB-113-0005 CAMB-113-0001
Bovine Ribonuclease A, DNase- and Protease-Free (from Pancreas)
Enzymes
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.
Applications include, purifying DNA plasmids, removing RNA from DNA, or any type of nucleic acid research.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty