Would you like to log in again?
Staphylococcus aureus V8 Protease
Supplier: Rockland Immunochemicals
MB-114-0001 MB-114-0001
RLMB-114-0001EA 174 USD InStock
RLMB-114-0001 CAMB-114-0001
Staphylococcus aureus V8 Protease
Enzymes
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.
Application: Cleaving peptide bonds on the COOH-terminal side of either aspartic or glutamic acids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty