Would you like to log in again?
Paenibacillus polymyxa Neutral Protease Dispase
Supplier: Rockland Immunochemicals
MB-127-0050 MB-127-0050
RLMB-127-0050EA 731 USD InStock
RLMB-127-0050 CAMB-127-0050
Paenibacillus polymyxa Neutral Protease Dispase
Enzymes
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.
Application: Separate skin epidermis from dermis leaving intact epithelial sheets and stem cell, hepatocyte and other cell isolation applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty