Would you like to log in again?
Diamond DNA Ladder 100 to 3000 bp, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-203-0500
RLMB-203-0500EA 118.31 USD InStock
RLMB-203-0500
Diamond DNA Ladder 100 to 3000 bp, Rockland Immunochemicals
Electrophoresis Reagents Molecular Weight Markers
Diamond DNA Ladder 100-3000bp
  • comprised of PCR products and double-stranded DNA
  • digested with restriction enzymes, phenol extracted, equilibrated to 10 mM Tris-HCl (pH 8.0) and 10mM EDTA
Diamond Ready-to-Use DNA Ladder 100-3000bp is a unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 12 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.54 μg a load) for approximating the mass of DNA in comparably intense samples of similar size.
Diamond Ready-to-Use DNA Ladder 100-3000bp has a suggested load volume is 5 µL/well.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty