Would you like to log in again?
Ruby Prestained Protein Standard 10 to 175 kDa, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-208-0500
RLMB-208-0500EA 222.05 USD InStock
RLMB-208-0500
Ruby Prestained Protein Standard 10 to 175 kDa, Rockland Immunochemicals
Electrophoresis Reagents Molecular Weight Markers
Ruby Prestained Protein Standard 10-175kDa
  • 11 proteins
  • 0.2-0.4 mg/ml of each protein in 20 mM Trisphosphate pH 7.5, 2% SDS, 1 mM 2-Mercaptoethanol, 3.6 M Urea, and 15% (v/v) glycerol
  • sharp, tight bands in the range of 10-175 kDa
Ruby Prestained Protein Standard 10-175kDa contains 11 proteins that resolve into sharp, tight bands in the range of 10-175 kDa. The standard allows you to monitor molecular weight separation during electrophoresis, estimate molecular weights of proteins of interest, and evaluate western blot transfer efficiency.
Ruby Prestained Protein Standard 10-175kDa is supplied in gel loading buffer and is ready to use. Do not heat, dilute, add reducing agent before loading. Suggested load volumes: 5 μl per well for mini-gels, 2.5 μl per well for blots - 10 μl per well for large gels, 5 μl per well for blots
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty