Would you like to log in again?
Revitablot™ Western Blot Stripping Buffer
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MB-085-0050 MB-085-0500
RLMB0850050EA 81 USD InStock
RLMB0850050 RLMB0850500
Revitablot™ Western Blot Stripping Buffer
Buffers Electrophoresis Buffers Stripping Buffers
Revitablot™ Western Blot Stripping Buffer
  • Ready-to-Use
  • Faster and efficient stripping of primary and secondary antibodies with low backgrounds
  • Gentle and effective on both nitrocellulose and PVDF membranes.
Revitablot™ Western Blot Stripping Buffer is a uniquely formulated, ready-to-use reagent specifically designed for western blotting. Revitablot™ Western Blot Stripping Buffer is faster and more efficient at stripping of primary and secondary antibodies, including HRP and fluorescent labeled. Blots can be stripped multiple times for the repeated use of membranes. This solution can be used at room temperature. Only incubation of 5-20 minutes needed to strip the membrane. Revitablot™ Western Blot Stripping Buffer is gentle and effective on both nitrocellulose and PVDF western blot membranes.
Application Note: Revitablot™ Western Blot Stripping Buffer is ready-to-use as a working 1X solution and requires no further dilution.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty