Would you like to log in again?
Mouse Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MS-T035 MS-T033 MS-T013 MS-T014 MS-T030 MS-T001 MS-T032 MS-T035 MS-T029 MS-T002 MS-T023 MS-T020 MS-T036 MS-T004 MS-T022 MS-T011 MS-T017 MS-T034 MS-T021 MS-T012 MS-T032 MS-T028 MS-T004 MS-T022 MS-T033 MS-T002 MS-T016 MS-T017 MS-T031 MS-T010 MS-T036 MS-T031 MS-T021 MS-T023 MS-T001 MS-T010 MS-T020 MS-T012 MS-T030 MS-T034 MS-T016 MS-T029 MS-T013 MS-T014 MS-T028 MS-T011
RLMS-T031EA 55 USD InStock
RLMS-T031 RLMS-T030 RLMS-T017 RLMS-T016 RLMS-T014 RLMS-T036 RLMS-T035 RLMS-T013 RLMS-T012 RLMS-T034 RLMS-T033 RLMS-T011 RLMS-T032 RLMS-T010 CAMS-T004 CAMS-T028 CAMS-T029 CAMS-T020 CAMS-T021 CAMS-T022 CAMS-T023 CAMS-T001 CAMS-T002 RLMS-T020 RLMS-T028 RLMS-T004 RLMS-T002 RLMS-T001 RLMS-T023 RLMS-T022 RLMS-T021 CAMS-T036 CAMS-T014 CAMS-T016 CAMS-T017 RLMS-T029 CAMS-T030 CAMS-T031 CAMS-T032 CAMS-T010 CAMS-T011 CAMS-T033 CAMS-T034 CAMS-T012 CAMS-T035 CAMS-T013
Mouse Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Use for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic-free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty