Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Peroxidase (PAP) Soluble Complex Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
P200-0005
RLP200-0005EA 175 USD InStock
RLP200-0005
Anti-Peroxidase (PAP) Soluble Complex Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Primary Mouse Anti-Peroxidase (PAP) Soluble Complex Reacts with N/A

Type: Primary
Antigen: Peroxidase (PAP) Soluble Complex
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype:
Reactivity: None

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty