Would you like to log in again?
Animal Whole Blood, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R300-0100 R109-0100 R305-0100 R302-0050 R312-0050 R310-0100 R310-0050 R317-0100 R304-0100 R204-0100 R105-0100 R109-0100 R302-0100 R305-0100 R105-0100 R308-0050 R300-0050 R104-0100 R308-0050 R307-0050 R311-0100 R310-0050 R310-0100 R210-0100 R202-0100 R111-0100 R313-0100 R109-0050 R217-0100 R211-0100 R213-0100 R311-0050 R300-0050 R309-0100 R307-0050 R306-0050 R306-0050 R200-0100 R309-0050 R207-0100 R311-0050 R311-0100 R107-0050 R309-0100 R111-0100 R105-0050 R305-0050 R302-0100 R100-0050 R111-0050 R100-0100 R111-0050 R205-0100 R102-0100 R307-0100 R209-0100 R109-0050 R309-0050 R214-0100 R206-0050 R307-0100 R113-0100 R107-0100 R212-0050 R302-0050 R105-0050 R305-0050
RLR107-0050EA 241.48 USD InStock
CAR310-0100 RLR107-0050 RLR317 CAR300-0050 RLR311-0100 RLR111-0100 RLR307-0050 CAR306-0050 RLR310-0100 RLR302-0100 RLR109-0100 RLR200 CAR302-0100 RLR204 CAR307-0050 RLR309-0100 RLR205 RLR202 RLR209 RLR206 RLR207 RLR308-0050 RLR105-0050 RLR305-0050 RLR310-0050 RLR302-0050 CAR107-0100 CAR307-0100 RLR211 RLR212 RLR210 RLR213 RLR214 RLR217 CAR109-0050 CAR309-0050 RLR300-0100 RLR305-0100 RLR100-0100 CAR311-0050 CAR111-0050 RLR105-0100 RLR102 RLR304 CAR305-0100 CAR105-0100 RLR104 RLR307-0100 CAR311-0100 CAR109-0100 CAR309-0100 RLR306-0050 CAR305-0050 CAR302-0050 CAR308-0050 RLR109-0050 RLR309-0050 CAR111-0100 CAR105-0050 RLR300-0050 RLR100-0050 CAR310-0050 RLR311-0050 RLR113 RLR111-0050 RLR312 RLR313
Animal Whole Blood, Rockland Immunochemicals
Blood
Use for DNA isolation
Blood is obtained from healthy, non-medicated animals. All blood, except hamster, mouse, pigeon, and rat blood is sterile.
Ordering information: For additional coagulants, sizes, or species, contact your Avantor representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty